Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
Código de producto. 30040270 Tienda Thermo Scientific Productos
Click to view available options
Capacidad (métrico):
50 L
100 L
200 L
Detalle del puerto:
4 x puertos y tubos de montaje superior
Puerto de polvo
Tamaño de la unidad:
paquete de 15
paquete de 3
paquete de 5
Código de producto. 30040270
Marca: Thermo Scientific™ SH30661.03
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
TRUSTED_SUSTAINABILITY
Especificaciones
Capacidad (métrico) 200 L
Detalle del puerto 4 x puertos y tubos de montaje superior
Línea de tuberías 1 0.25 in. ID Tubing and male/female Luer connectors (Port 1)
Línea de tuberías 2 0.25 in. ID Tubing and male/female Luer connectors (Port 2)
Línea de tuberías 3 3/8 in. ID tubing and quick-connect couplings (Port 3)
Línea de tuberías 4 3/8 in. ID tubing and quick-connect couplings (Port 4)
Tubing Style TPE
Esterilidad Sterile