missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIXL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA564903
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: RKRTSFSAEQL QLLELVFRRT RYPDIHLRER LAALTLLPES RIQVWFQNRR AKSRRQSGKS FQPLARPEII LNHCAPGTET KCLKPQLPLE VDVNCLPEPN GVGGGISDSS SQGQNFETCS PLSEDIGSKL DSWEEHIFSA Highest antigen sequence identity to the following orthologs - mouse 79%, rat 79%.
MIXL1 is a novel human Mix-like homeobox gene. In normal hematopoiesis, its expression appears to be restricted to immature B- and T-lymphoid cells.
Especificaciones
| MIXL1 | |
| Polyclonal | |
| Unconjugated | |
| MIXL1 | |
| hMix; Homeobox protein MIXL1; homeodomain protein MIX; MILD1; MIX; Mix paired-like homeobox; Mix paired-like homeobox 1; mix.1 homeobox-like protein; Mix1 homeobox-like 1 (Xenopus laevis); MIX1 homeobox-like protein 1; MIXL; Mixl1; Mix-like homeobox protein 1; Mm1; mMix; Mml | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83881 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2W2 | |
| MIXL1 | |
| Recombinant Human MIXL1. Recombinant protein control fragment (Product #RP-105701). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido