missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIP-3 alpha Monoclonal Antibody (114908)
Rat Monoclonal Antibody
Marca: Invitrogen™ MA523857
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody does not cross-react with recombinant human (rh) CCL19, rhCCL20, rmCCL3, 4, 9, 19, rmCXCL2, or rrCCL20. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
Especificaciones
| MIP-3 alpha | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O89093 | |
| CCL20 | |
| E. coli-derived recombinant mouse CCL20/MIP-3 alpha Ala28-Met97. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Neutralization | |
| 114908 | |
| Unconjugated | |
| CCL20 | |
| beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 α; MIP 3alpha; MIP 3α; MIP3 a; MIP3 alpha; MIP3 α; MIP-3[a]; MIP3A; MIP-3A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20297 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido