missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MINT3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 30533961
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30533961 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30533961 Proveedor Invitrogen™ N.º de proveedor PA5149047

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

The munc-18 interacting protein (Mint) protein family is a group of evolutionarily conserved adaptor proteins that function in membrane transport and organization. In mammals, there exist three mint isoforms, Mint1, 2, and 3. Although there is little amino acid sequence conservation in the amino-terminal half, the carboxy-terminal half of these proteins is highly conserved. Within this conserved portion there exists a phosphotyrosine-binding (PTB) and a PSD-95/DLG-A/ZO-1 (PDZ) domain, which function as protein interaction modules. Mint1 and 2 appear to be expressed exclusively in the brain and are found to bind to Munc18, an essential component of the synaptic vesicle fusion machinery. Mint3 is ubiquitously expressed in all tissues and is expressed at the lowest levels in the brain and testis. Studies show that mint3 does not interact with munc-18. Mint3 has been found to interact with the Alzheimer's Disease-related amyloid precursor protein (APP) and does so through its PTB and PDZ domains. It has been suggested that mint3 links APP to other transport machinery components, thereby regulating it transport, endocytosis, and metabolism. Abnormal APP metabolism has been shown to be the cause of an early-onset type of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MINT3
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with trehalose and no preservative
génica APBA3
N.º de referencia del gen O96018
Alias de gen Adapter protein X11gamma; amyloid beta (A4) precursor protein-binding family A member 3 (X11-like 2); amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2); amyloid beta (A4) precursor protein-binding, family A, member 3; amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2); amyloid beta A4 precursor protein-binding family A member 3; amyloid beta precursor protein binding family A member 3; Amyloid-beta A4 precursor protein-binding family A member 3; Apba3; AW208791; lin 10; lin-10; MGC:15815; Mint 3; MINT3; mint-3; Neuronal Munc18-1-interacting protein 3; neuron-specific X11L2 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; X11gamma; X11L2; X11-like 2 protein
Símbolos de los genes APBA3
Especie del huésped Sheep
Inmunógeno E. coli-derived recombinant human APBA3, Met1-Leu138.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9546
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.