Learn More
Milbemycin oxime, MedChemExpress
Código de producto. 16411446
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
200 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
200 miligramos
50 miligramos
16411446 200 mg 200 miligramos
16458608 10 mg 10 miligramos
16428456 100 mg 100 miligramos
16404487 50 mg 50 miligramos
4 options
Código de producto. 16411446
Marca: Medchem Express HYB0778
MedChemExpress Milbemycin oxime is a macrocyclic lactone and has broad-spectrum anti-parasitic activity. Milbemycin oxime is composed of milbemycins A4 and A3. Milbemycin oxime binds glutamate-gated chloride channels. Milbemycin oxime is against intestinal nematodes, pulmonary and cardiac helminths.
Milbemycin oxime is a macrocyclic lactone and has broad-spectrum anti-parasitic activity. Milbemycin oxime is composed of milbemycins A4 and A3. Milbemycin oxime binds glutamate-gated chloride channels. Milbemycin oxime is against intestinal nematodes, pulmonary and cardiac helminths.
Especificaciones
Nombre del producto químico o material Milbemycin oxime
Forma física Solid
CAS 129496-10-2
Color Blanco
Cantidad 200 mg
Notas de grado de pureza Research
Fórmula molecular C63H88N2O14
Para utilizar con (aplicación) COVID-19-immunoregulation
Información de solubilidad DMSO : ≥ 100 mg/mL (91.13 mM) ∣H2O : 1 mg/mL (0.91 mM; ultrasonic and warming and heat to 80°C)
SMILES CC1=C[C@@H]2[C@@]3(O)[C@H](OC/C3=C\C=C\[C@H](C)C/C(C)=C/C[C@@H](O[C@@]4(CC[C@H](C)[C@@H](C)O4)C5)C[C@@H]5OC2=O)/C1=N\O.CC6=C[C@@H]7[C@@]8(O)[C@H](OC/C8=C\C=C\[C@H](C)C/C(C)=C/C[C@@H](O[C@@]9(CC[C@H](C)[C@@H](CC)O9)C%10)C[C@@H]%10OC7=O)/C6=N\O
Peso molecular (g/mol) 1097.38
Formula Weight (peso de la fórmula) 1097.38
Porcentaje de pureza 99.45%