Learn More
Heinz Herenz Microscope Slides with Cellophane
Click to view available options
:
Not Frosted
Descripción
We offer a very wide range of slides, so you are sure to find what you are looking for.
- Within the product lines you will find cut and ground slides.
- Cut slides are preferred for routine microscopy and offer great flexibility.
- Ground slides prevent cuts due to the ground edges.
- If you want to label and categorize your slides, it is best to choose slides with matted edges or a coloured field.
- You also have the choice between slides with and without cellophane.
Especificaciones
Especificaciones
| Esterilidad | Non-sterile |
| Edges | Cut |
| Frosted | Not Frosted |
| Material | Glass |
| Precleaned | Yes |
| Tipo | Plain Microscope Slide |
| Número de pocillos | 0 |
| Color | Clear |
| Estilo de esquina | Plain |