Learn More
Excelon Micro-Line Translucent Mini-Bore Tubing
Código de producto. p-200105127 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
0,02 in
0,03 in
0,04 in
0,05 in
0,063 in
Diámetro exterior (imperial):
0,06 in
0,07 in
0,09 in
0/125 in
Tamaño de la unidad:
15 metros
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30053157 0,02 in 0,06 in 15 metros
30054321 0,05 in 0,09 in 15 metros
30053547 0,04 in 0,07 in 15 metros
30054063 0,03 in 0,09 in 15 metros
30053014 0,063 in 0/125 in 15 metros
5 options
Código de producto. 30053157
Marca: Excelon 5011750
Solo quedan null
Añadir a la cesta
Añadir a la cesta
A translucent, odorless, and tasteless mini-bore tubing that offers practical solutions to maintaining a non-toxic and non-contaminating environment
Meets USP Class VI, FDA Regulation 21CFR177.350, and National Formulary Vo. 14. No plasticizers used in the manufacturing process. Excellent flexibility with no kinking and 100% plastic memory. Retains flexibility in sub-zero temperatures and resists age hardening. Won’t leach or migrate
Especificaciones
Diámetro interior (imperial) 0,02 in
Diámetro exterior (imperial) 0,06 in
Color Translúcido
Tipo de producto Micro-Line Tubing