missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA524865
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with bovine based on sequence homology.
MFAP4 is a protein with similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region.
Especificaciones
| MFAP4 | |
| Polyclonal | |
| Unconjugated | |
| MFAP4 | |
| 1110007F23Rik; 36 kDa MAP; 36 kDa microfibril-associated glycoprotein; Magp-36; MFAP4; microfibril associated protein 4; microfibril-associated glycoprotein 4; microfibrillar associated protein 4; microfibrillar-associated protein 4; Unknown (protein for MGC:137920) | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 4239, 76293 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P55083, Q9D1H9 | |
| MFAP4 | |
| KLH conjugated synthetic peptide between 228-255 amino acids from the C-terminal region of human MFAP4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido