Learn More
Thermo Scientific Chemicals Metilcelulosa, viscosidad 8000 cps
Descripción
Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. It is also used as binders, lubricants, steric stabilizer, water retention aid and sizing in the production of papers and textiles as it protects the fibers from absorbing water or oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Methyl cellulose |
| Nombre de nota | Viscosity: 8000cPs |
| Olor | Odorless |
| Cantidad | 100 g |
| Índice Merck | 14,6040 |
| Información de solubilidad | Soluble in glacial acetic acid and cold water. Insoluble in hot water,ethanol,ether and chloroform. |
| Forma física | Polvo |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.