Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-480
Click to view available options
Tamaño (métrico):
101,6 mm x 50,8 mm (An x Al)
101,6 mm x 91.4 mm (An x Al)
19,05 × 6,35 mm (An × Al)
25,4 x 9,52 mm (An x Al)
38,1 x 12,7 mm (An x Al)
38,1 × 19,05 mm (An × Al)
50,8 x 25,4 mm (An x Al)
50,8 mm x 12,7 mm (An x Al)
50,8 mm x 91,4 mm (An x Al)
69,85 mm x 31,75 mm (An x Al)
76,2 mm x 25,4 mm (An x Al)
76,2 mm x 50,8 mm (An x Al)
76,2 mm x 91,4 mm (An x Al)
Tamaño de la unidad:
1 rollo
Descripción
- Available in various sizes
- Rubber-based Mondo Bondo™ adhesive for oily, greasy applications
Especificaciones
Especificaciones
| Certificaciones/conformidad | UL Recognised; cUL Recognised |
| Descripción | Thermal Transfer Printable Labels |
| Tipo de adhesivo | Permanent Rubber |
| Recubrimiento | None |
| Color | Light Gray |
| Acabado | Satin |
| Formato | Rollo |
| Material | Polyester (B-480) |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Mostrar más |