Learn More
GA International Metal Rack Labels for Barcode and Thermal Printers on Small Roll
Descripción
- Specifically designed for the identification of metal and stainless-steel surfaces
- Resistant to extreme cryogenic conditions, including liquid nitrogen (-196°C) and ultra-low temperature freezers
- Printable with all major brands of barcode printers
- Stored on a small roll with core size of 25.4 mm
Especificaciones
Especificaciones
| Tipo de adhesivo | Permanent |
| Color | Naranja |
| Acabado | Gloss |
| Formato | Rollo |
| Material | Thermoplastic Film |
| Print Technology | Thermal Transfer |
| Tipo de producto | Label |
| Esterilidad | No estéril |
| Etiquetas por fila | 1 Label |
| Cantidad | 2000/Pk. |
| Mostrar más |