missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meta-Pneumovirus Antibody (HMPV57), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP1-25941JF646
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones
| Meta-Pneumovirus | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GLYG | |
| Sub-type A hMPV virus isolate NCL03-4/174 | |
| 0.1 mL | |
| 2799942 | |
| Virus | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| HMPV57 | |
| Immunocytochemistry/Immunofluorescence | |
| hMPV, metapneumovirus | |
| Mouse | |
| Protein G purified | |
| Primary | |
| NBP1-25941 recognises human metapneumovirus (hMPV), a member of the Paramyxoviridae RNA virus family. Metapneumovirus was discovered in 2001 and is distributed worldwide. Infection in the upper respiratory tract is associated with mild illness, while lower respiratory tract infection can cause fever, cough, pneumonia, bronchiolitis and wheezing. Severe lower respiratory tract infection is most common in the very young, the very old and the immunosuppressed. Metapneumovirus is a common cause of respiratory infection.Phylogenetic analysis has shown two major genotypes, A and B, each containing two sub-divisions, suggesting that four distinct lineages circulate in the population with different lineages predominating in different years. NBP1-25941 recognises all sub-types (A1, A2, B1 and B2) of hMPV NBP1-25941 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria.The target antigen has not been determined for this antibody. | |
| Store at 4°C in the dark. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido