Learn More
Melford MES Sodium Salt
Descripción
MES Sodium Salt, also known as 2-(N-Morpholino)ethanesulfonic acid sodium salt, is a zwitterionic buffer commonly used in biological and biochemical research.
In the life science field, MES Sodium Salt is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 5.5 to 6.7, making it suitable for applications requiring slightly acidic to neutral pH conditions.
MES Sodium Salt is commonly used in protein purification, enzyme assays, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.
One of the significant advantages of MES Sodium Salt is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.
Especificaciones
Especificaciones
| Nombre del producto químico o material | MES Sodium Salt |
| CAS | 71119-23-8 |
| Cantidad | 500 g |
| Fórmula molecular | C6H12NNaO4S |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.