missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Spike RBD MAb (Cl 1038342), R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB107071-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Spike RBD Monoclonal antibody specifically detects Spike RBD in MERS-CoV samples. It is validated for Blocking Assay, Immunocytochemistry
Especificaciones
Spike RBD | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Blocking Assay, Immunocytochemistry | |
1038342 | |
Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL | |
YP_007188579.1 | |
Chinese Hamster Ovary cell line CHO-derived MERS-CoV Spike RBD, Glu367-Tyr606, Accession # YP_007188579.1 | |
25 μg | |
Primary | |
MERS-CoV | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
MERS-CoV Spike RBD MAb (Cl 1038342), R&D Systems™ > 25 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido