Learn More
MEP1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA525752
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
Especificaciones
| MEP1A | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| P28825, Q16819 | |
| MEP1A | |
| KLH conjugated synthetic peptide between 149-175 amino acids from the N-terminal region of human MEP1A | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| MEP1A | |
| AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; endopeptidase-24.18 subunit alpha; Mep1; Mep-1; MEP1A; Mep-1a; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 17287, 4224 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |