missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody

Código de producto. 12671187 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
500 μL
1 mL
7 mL
12 mL
Formulario:
Concentrated
Prediluted
Ready-to-Use
Tamaño de la unidad:
1 ml
100 microlitros
12 ml
500 microlitros
7 ml
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Formulario unitSize
12671187 12 mL Ready-to-Use 12 ml
12691187 100 μL Concentrated 100 microlitros
12601197 500 μL Concentrated 500 microlitros
12681187 1 mL Concentrated 1 ml
12661187 7 mL Prediluted 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 12671187 missing translation for 'mfr' Epredia™ missing translation for 'supplierNo' MS364RQ

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available wit the Epredia™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody.

Research Areas: Cancer Marker

Host Species: Mouse

Clone: HMB45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with dog and rat. Others not known.

Immunogen: Extract of pigmented melanoma metastases from lymph nodes

Molecular Weight: 100kDa

Positive Control: Melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

    Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antígeno Melanoma (gp100) Ab-1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon HMB45
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Extract of pigmented melanoma metastases from lymph nodes
Cantidad 12 mL
Estado normativo IVD
Disciplina de investigación Cancer and Tumor Biology
Primario o secundario Primary
Formulario Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.