missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MED14 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16546271
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
16546271 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16546271

Marca: Invitrogen™ PA587913

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: LDQATLDDME KSVNDDMKRI IPWIQQLKFW LGQQRCKQSI KHLPTISSET LQLSNYSTHP IGNLSKNKLF IKLTRLPQYY IVVEMLEVPN KPTQLSYKYY FMSVNAADRE DSPAMALLLQ QFKENIQDLV FRTKTGKQTR TNAKRKLSDD PCPVESKKTK RAGEMCAFNK VLAHFVAMCD T; Positive Samples: HeLa, SW480, Jurkat, Mouse brain, Mouse pancreas; Cellular Location: Nucleus.

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MED14
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 3.54 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica Med14
N.º de referencia del gen A2ABV5, O60244
Alias de gen 9930001L01Rik; Activator-recruited cofactor 150 kDa component; ARC150; AU041628; Cofactor required for Sp1 transcriptional activation subunit 2; cofactor required for Sp1 transcriptional activation subunit 2 (150 kDa); cofactor required for Sp1 transcriptional activation, subunit 2; cofactor required for Sp1 transcriptional activation, subunit 2 (150kD); CRSP complex subunit 2; CRSP150; CRSP2; CSRP; CXorf4; DRIP150; ENSMUSG00000073278; EXLM1; Gm641; hRGR1; human homolog of yeast RGR1; MED14; Mediator complex subunit 14; mediator of RNA polymerase II transcription subunit 14; MGC104513; ORF1; RGD1560170; RGR1; RGR1 homolog; thyroid hormone receptor-associated protein complex 170 kDa component; thyroid hormone receptor-associated protein complex component TRAP170; Transcriptional coactivator CRSP150; transcriptional co-activator CRSP150; TRAP170; vitamin D receptor-interacting protein complex component DRIP150; vitamin D3 receptor-interacting protein complex 150 kDa component
Símbolos de los genes Med14
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 470-650 of human MED14 (NP_0042202).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 26896, 317343, 9282
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.