Learn More
Invitrogen™ MDA5 Monoclonal Antibody (K49007_11D6), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA555914
Descripción
Sequence of this protein is as follows: QIQEPCKKFA IADATREDPF KEKLLEIMTR IQTYCQMSPM SDFGTQPYEQ WAIQMEKKAA KEGNRKERVC AEHLRKYNEA LQINDTIRMI DAYTHLETFY NEEKDKKFAV IEDDSDEGGD DEYCDGDEDE DDLKKPLKLD ETDRFLMTLF FENNKMLKRL AENPEYENEK LTKLRNTIME QYTRTEESAR GIIFTKTRQS AYALSQWITE NEKFAEVGVK AHHLIGAGHS SEFKPMTQNE QKEVISKFRT GKINLLIATT VAEEGLDIKE CNIVIRYGLV TNEIAMVQAR
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein. Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.
Especificaciones
| MDA5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9BYX4 | |
| IFIH1 | |
| Recombinant human IFIH1 (immunogen range is 531-820). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K49007_11D6 | |
| Unconjugated | |
| IFIH1 | |
| 9130009C22Rik; AGS7; CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; helicard; Helicase with 2 CARD domains; Hlcd; IDDM19; IFIH1; interferon induced with helicase C domain 1; interferon induced with helicase C domain protein 1; interferon induced, with helicase C domain 1; interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA5; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RGD1560496; RH116; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116; SGMRT1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 64135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.