missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCT2 Polyclonal Antibody
Goat Polyclonal Antibody
Marca: Invitrogen™ PA517960
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Tissues with few or no mitochondria, such as erythrocytes and tumor cells, depend largely on glycolysis to generate ATP. The major end products of glycolysis, pyruvate and lactate, must be eliminated from these cells to enable continued glycolytic flux and prevent toxic effects. H+/monocarboxylate transporters mediate the transport of lactate and pyruvate. Human MCT2 has a high affinity for the transport of pyruvate.
Especificaciones
| MCT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A7 | |
| 4921534N07Rik; MCT 2; Mct2; monocarboxylate transporter; Monocarboxylate transporter 2; Slc16a7; solute carrier family 16 (monocarboxylate transporter), member 7; solute carrier family 16 (monocarboxylic acid transporters), member 7; solute carrier family 16 member 7; solute carrier family 16, member 7 (monocarboxylic acid transporter 2) | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9194 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60669 | |
| SLC16A7 | |
| Synthetic peptide sequence (SNAQSVTSERETNI) corresponding to the C-terminus amino acids of SLC16A7 (aa 465-478). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido