Learn More
MCP-5 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Descripción
AA Sequence of recombinant protein: GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-5 (CCL12). Anti-Murine MCP-5 (CCL12)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mMCP-5 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine MCP-5 (500-P61) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mMCP-5. Western Blot: To detect mMCP-5 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mMCP-5 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P61BT-1MG will be provided as 2 x 500 μg
Especificaciones
Especificaciones
| Antígeno | MCP-5 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with no preservative |
| génica | Ccl12 |
| N.º de referencia del gen | Q62401 |
| Alias de gen | C-C motif chemokine 12; Ccl12; chemokine (C-C motif) ligand 12; MCP-1-related chemokine; Mcp5; MCP-5; Monocyte chemoattractant protein 5; monocyte chemotactic protein 5; Scya12; small inducible cytokine A12; small-inducible cytokine A12 |
| Símbolos de los genes | Ccl12 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.