missing translation for 'onlineSavingsMsg'
Learn More

PeproTech MCP-1 Polyclonal Antibody, PeproTech®, Invitrogen™

Código de producto. 17883483
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17883483 50 μg 50 microgramos
17843173 100 μg 100 microgramos
17873253 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17883483

Marca: PeproTech 500P7650UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: QPDAVNAPLT CCYSFTGKMI PMSRLENYKR ITSSRCPKEA VVFVTKLKRE ICADPNKEWV QKYIRKLDQN QVRSETTVFY KIASTLRTSA PLNVNLTHKS EANASTLFST TTSSTSVEVT SMTEN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat MCP-1 (CCL2). Anti-Rat MCP-1 (CCL2)-specific antibody was purified by affinity chromatography employing an immobilized Rat MCP-1 (CCL2) matrix. Sandwich ELISA: To detect Rat MCP-1 (CCL2) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat MCP-1 (CCL2) (500-P76Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat MCP-1 (CCL2). Western Blot: To detect Rat MCP-1(MCAF) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat MCP-1(MCAF) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MCP-1
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica Ccl2
N.º de referencia del gen P14844
Alias de gen Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
Símbolos de los genes Ccl2
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Rat MCP-1 (CCL2).
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 24770
Especies diana Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.