Learn More
MCEE Monoclonal Antibody (OTI13B9), TrueMAB™, OriGene
Código de producto. 17569983
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17569983 30 μL 30 microlitros
17549983 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17569983 Proveedor Origene Technologies N.º de proveedor TA808575S
Mouse Monoclonal Antibody
MCEE Monoclonal antibody specifically detects MCEE in Human samples. It is validated for Western Blot
Especificaciones
Antígeno MCEE
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI13B9
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica AMTN
N.º de referencia del gen Q6UX39
Alias de gen 1110007A04Rik; DL-methylmalonyl-CoA racemase; GLOD2; glyoxalase domain containing 2; Mcee; methylmalonyl CoA epimerase; methylmalonyl-CoA epimerase; methylmalonyl-CoA epimerase, mitochondrial
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human AMTN produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 401138
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG2a