Learn More
Termómetro de inmersión total de alcohol azul MBL™
Filled with safe low toxicity blue or green indicating fluid and calibrated for total immersion. MBL™ Blue Spirit Total Immersion Thermometer gives accurate readings when the thermometer is immersed to a specific depth. Graduations are marked in permanent amber stain. White background provides clear observation of colored liquid column. Reinforced bulb allows stirring. Includes plastic non-roll top and safety suspension ring.
Marca: Pyrex 30/110/10
Descripción
- Mercury free - filled with a low toxicity blue or green spirit
- Calibrated for total immersion
- Permanent amber stain graduations
- White colored background provides clear observation of blue colored liquid column
- Reinforced bulb for stirring
- With plastic non-roll top and safety suspension ring
- Nominal length 305mm with item 30/210/10 length 405mm