missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).
Specifications
Specifications
| Antígeno | MBD5 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.26 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | MBD5 |
| N.º de referencia del gen | B1AYB6, Q9P267 |
| Alias de gen | KIAA1461, MBD5, MRD1 |
| Símbolos de los genes | MBD5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?