missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD4 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-91189F
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MBD4 Polyclonal specifically detects MBD4 in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Especificaciones
| MBD4 | |
| Polyclonal | |
| Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative) | |
| O95243 | |
| MBD4 | |
| A synthetic peptide made to an internal portion of the human MBD4 protein (between residues 200-250) [UniProt O95243] | |
| 0.1 mL | |
| Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics | |
| 8930 | |
| Human, Mouse (Negative) | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Does not react with mouse. | |
| Store at 4C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido