Learn More
BOLA Maxi Propeller Blades
Código de producto. p-200109994
Change view
Click to view available options
Dimensiones (Largo x Ancho x Alto):
140 x 20 x 4 mm
200 x 20 x 6 mm
200 x 26 x 6 mm
Tamaño de la unidad:
cada uno
Código de producto. Dimensiones (Largo x Ancho x Alto) unitSize
10080772 140 x 20 x 4 mm cada uno
16229621 200 x 20 x 6 mm cada uno
10326152 200 x 26 x 6 mm cada uno
3 options
Código de producto. 10080772
Marca: BOLA C44110
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Maxi Propeller Blades for stainless steel shafts jacketed with PTFE, glass shafts or metal shafts
  • These solid stirrer blades are made of PTFE and have a set of clamp screws made of a PTFE/PEEK compound.
  • The blades can be fixed tightly on BOLA Stirrer Shafts by means of the clamp screws. A spanner wrench is included for easy assembly.
Especificaciones
Dimensiones (Largo x Ancho x Alto) 140 x 20 x 4 mm
Recubrimiento PTFE