missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MATK Polyclonal Antibody

Descripción
Immunogen sequence: PEALKHGKFT SKSDVWSFGV LLWEVFSYGR APYPKMSLKE VSEAVEKGYR MEPPEGCPGP VHVLMSSCWE AEPARRPPFR KLAEKLAREL RSAGAPASVS GQDADGSTSP RSQEP; Positive Samples: THP-1; Cellular Location: Cytoplasm, Membrane.
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
Especificaciones
Especificaciones
| Antígeno | MATK |
| Aplicaciones | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.29 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | MATK |
| N.º de referencia del gen | P41242, P41243, P42679 |
| Alias de gen | Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase |
| Símbolos de los genes | MATK |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?