Learn More
MAT1A Rabbit anti-Human, Mouse, Plant, Rat, Polyclonal, Proteintech
Código de producto. 16880094 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16880094 150 μL 150 microlitros
16890094 20 μL 20 microlitros
2 options
Código de producto. 16880094
Marca: Proteintech 123951AP150UL
Rabbit Polyclonal Antibody
This antibody was raised against the full length of human MAT1A and may cross-react with MAT2A. Western blot analysis using this antibody detected a single band around 44-50kD in liver.
MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.
Especificaciones
Antígeno MAT1A
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica MAT1A
N.º de referencia del gen P13444, Q00266, Q91X83
Alias de gen AdoMet synthase 1, AMS1, MAT, MAT 1, MAT I/III, MAT1A, MATA1, SAMS, SAMS1
Símbolos de los genes MAT1A
Especie del huésped Rabbit
Inmunógeno MAT1A Fusion Protein Ag3076
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11720, 25331, 4143
Especies diana Human, Mouse, Plant, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG