missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH6, Mouse anti-Human, Clone: 1A5, Abnova™
Descripción
Sequence: DTAEEDICRVCRSEGTPEKPLYHPCVCTGSIKFIHQECLVQWLKHSRKEYCELCKHRFAFTPIYSPDMPSRLPIQDIFAGLVTSIGTAIR
Especificaciones
Especificaciones
| Antígeno | MARCH6 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1A5 |
| Conjugado | Unconjugated |
| Formulación | In 1x PBS, pH 7.4 |
| génica | membrane-associated ring finger (C3HC4) 6 |
| N.º de referencia del gen | NM_005885 |
| Alias de gen | KIAA0597/MARCH-VI/RNF176/TEB4 |
| Símbolos de los genes | MARCH6 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?