missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MARCH1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15856593
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Product Code. Cantidad unitSize
15856593 100 μL 100 microlitros
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 15856593

missing translation for 'mfr': Invitrogen™ PA568179

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MARCH1 Polyclonal Antibody for Western Blot, IHC (P)

E3 ubiquitin-protein ligase that mediates ubiquitination of TFRC, CD86, FAS and MHC class II proteins, such as HLA-DR alpha and beta, and promotes their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. By constitutively ubiquitinating MHC class II proteins in immature dendritic cells, down-regulates their cell surface localization thus sequestering them in the intracellular endosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno MARCH1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica MARCH1
N.º de referencia del gen Q8TCQ1
Alias de gen 1-Mar; 2900024D24Rik; BB085186; E3 ubiquitin-protein ligase MARCH1; MARCH1; MARCHF1; MARCH-I; membrane associated ring finger 1; membrane associated ring-CH-type finger 1; membrane-associated ring finger (C3HC4) 1; membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase; membrane-associated RING finger protein 1; Membrane-associated RING-CH protein I; RGD1305148; RING finger protein 171; RING-type E3 ubiquitin transferase MARCH1; RNF171
Símbolos de los genes MARCH1
Especie del huésped Rabbit
Inmunógeno Synthesized peptide derived from human MARCH1.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55016
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.