Learn More
MAPRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10364-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.Especificaciones
| MAPRE2 | |
| Polyclonal | |
| Unconjugated | |
| MAPRE2 | |
| APC binding protein EB2, EB1, EB2, End binding protein 2, MAPRE2, RP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10982, 212307, 679221 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15555, Q3B8Q0, Q8R001 | |
| Mapre2 | |
| MAPRE2 Fusion Protein Ag0416 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |