Learn More
MAPRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16856703 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16856703 150 μL 150 microlitros
16866703 20 μL 20 microlitros
2 options
Código de producto. 16856703
Marca: Proteintech 103641AP150UL
Rabbit Polyclonal Antibody
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.
Especificaciones
Antígeno MAPRE2
Aplicaciones Immunocytochemistry, Immunofluorescence, Western Blot
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica MAPRE2
N.º de referencia del gen Q15555, Q3B8Q0, Q8R001
Alias de gen APC binding protein EB2, EB1, EB2, End binding protein 2, MAPRE2, RP1
Símbolos de los genes Mapre2
Especie del huésped Rabbit
Inmunógeno MAPRE2 Fusion Protein Ag0416
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10982, 212307, 679221
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG