missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK2 Antibody (7H4.2) [Janelia Fluor™ 669], Novus Biologicals™
Descripción
MAPKAPK2 Monoclonal antibody specifically detects MAPKAPK2 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Especificaciones
Specifications
| Antígeno | MAPKAPK2 |
| Aplicaciones | Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | 7H4.2 |
| Conjugado | Janelia Fluor 669 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 |
| Especie del huésped | Mouse |
| Inmunógeno | KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?