missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK4 Monoclonal Antibody (OTI2F4)

Descripción
MAPK4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.
Especificaciones
Especificaciones
| Antígeno | MAPK4 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | OTI2F4 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | MAPK4 |
| N.º de referencia del gen | P31152 |
| Alias de gen | A330097D03Rik; Erk3; Erk3-related; ERK4; ERK-4; Extracellular signal-regulated kinase 4; MAP kinase 4; MAP kinase isoform p63; MAPK 4; MAPK4; mitogen activated protein kinase 4; mitogen-activated protein kinase 4; MNK2; p63Mapk; p63-MAPK; Prkm4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?