Learn More
MAPK13 Mouse anti-Human, Clone: OTI12D2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF505804
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and cellular stress. MAP kinase kinases 3, and 6 can phosphorylate and activate this kinase. Transcription factor ATF2, and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase.Especificaciones
| MAPK13 | |
| Monoclonal | |
| Unconjugated | |
| MAPK13 | |
| MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5603 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI12D2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O15264 | |
| MAPK13 | |
| Full length human recombit protein of human MAPK13 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.