Learn More
MAP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16891664 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16891664 150 μL 150 microlitros
16801674 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16891664 Proveedor Proteintech N.º de proveedor 134461AP150UL
Rabbit Polyclonal Antibody
The product of this gene is a microtubule-associated protein that is predomitly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Antígeno MAP7
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica MAP7
N.º de referencia del gen O88735, Q14244
Alias de gen E MAP 115, EMAP115, Ensconsin, MAP 7, MAP7
Símbolos de los genes Map7
Especie del huésped Rabbit
Inmunógeno MAP7 Fusion Protein Ag4276
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17761, 293016, 9053
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG