missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP4K5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA540649
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: QGKSFKSDEV TQEISDETRV FRLLGSDRVV VLESRPTENP TAHSNLYILA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 77%; Zebrafish: 86%.
MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases and has been implicated as an upstream regulator of MAP kinase signaling pathways. Yeast SPS1/STE20 functions near the beginning of the MAP kinase signal cascades that is essential for yeast pheromone response. This kinase was shown to activate Jun kinase in mammalian cells, which suggested a role in stress response. Two alternatively spliced transcript variants encoding the same protein have been described for MAP4K5.
Especificaciones
| MAP4K5 | |
| Polyclonal | |
| Unconjugated | |
| Map4k5 | |
| 4432415E19Rik; fl74c10; GCKR; germinal center kinase-related; KHS; KHS1; Kinase homologous to SPS1/STE20; LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 5; map4k5; MAPK/ERK kinase kinase kinase 5; MAPKKKK5; MEK kinase kinase 5; MEKKK 5; mitogen-activated protein kinase kinase kinase kinase 5; RGD1562028; wu:fl74c10; zgc:55719; zgc:55985 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11183 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y4K4 | |
| Map4k5 | |
| Synthetic peptide directed towards the middle region of human MAP4K5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido