missing translation for 'onlineSavingsMsg'
Learn More

MAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Código de producto. 16858163 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16858163 20 μL 20 microlitros
16848163 150 μL 150 microlitros
2 options
This item is not returnable. View return policy

Product Code. 16858163

Brand: Proteintech 112291AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it has been shown to play a role in the regulation of embryonic and ocular development. Mutations in this gene cause various glaucoma phenotypes including primary congenital glaucoma, autosomal domit iridogoniodysgenesis anomaly, and Axenfeld-Rieger anomaly.

Specifications

Antígeno MAP4
Aplicaciones Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica MAP4
N.º de referencia del gen P27546, P27816, Q5M7W5
Alias de gen MAP 4, MAP4
Símbolos de los genes Map4
Especie del huésped Rabbit
Inmunógeno MAP4 Fusion Protein Ag1741
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17758, 367171, 4134
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.