missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21610-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.Especificaciones
| MAP3K4 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K4 | |
| KIAA0213, MAP three kinase 1, MAP3K4, MAPK/ERK kinase kinase 4, MAPKKK4, MEK kinase 4, MEKK 4, MEKK4, MTK1, PRO0412 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26407, 308106, 4216 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08648, Q9Y6R4 | |
| Map3k4 | |
| MAP3K4 Fusion Protein Ag16254 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido