Descripción
The MAP2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human MAP2. This antibody recognizes human, mouse, rat antigen. The MAP2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
Especificaciones
| Antígeno | MAP2 |
| Aplicaciones | Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | CoraLite 488 |
| Especie del huésped | Rabbit |
| Cantidad | 100 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 4133 |
| Especies diana | Human, Mouse, Rat |
| Isotype | IgG |