missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP2 Monoclonal Antibody (HM-2)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA125043
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
Especificaciones
| MAP2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P11137, P15146, P20357 | |
| Map2 | |
| Rat brain microtubule-associated proteins (MAPs). | |
| 50 μL | |
| Primary | |
| Avian, Bovine, Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HM-2 | |
| Unconjugated | |
| Map2 | |
| a730034c02; AC091465.1; DKFZp686I2148; G1-397-34; MAP 2; Map2; MAP-2; map2.S; map2a; map-2a; map2ab; MAP2B; MAP2C; MAP2R; microtubule associated protein 2; microtubule associated protein 2 S homeolog; microtubule associated protein 2; microtubule-associated protein 2; microtubule associated protein 2C; microtubule-associated protein 2; microtubule-associated protein-2; Mtap2; Mtap-2; OTTHUMP00000163916; OTTHUMP00000208143; repro4; XELAEV_18047318mg; xmap2 | |
| Mouse | |
| Purified | |
| RUO | |
| 17756, 25595, 281294, 4133, 424001 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido