missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA316820
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is not recommended for ICC/IF applications.
The small PDZK1-associated protein (SPAP; also known as MAP17) is an endogenous regulator of cellular PDZK1 levels by regulating PDZK1 turnover.
Especificaciones
| MAP17 | |
| Polyclonal | |
| Unconjugated | |
| PDZK1IP1 | |
| 0610007F13Rik; 17 kDa membrane-associated protein; 2700030M23Rik; AU046042; DD96; epithelial protein up-regulated in carcinoma, membrane associated protein 17; MAP17; membrane-associated protein 17; membrane-associated protein 17 (Map17); PDZK1 interacting protein 1; PDZK1-interacting protein 1; PDZK1IP1; Protein DD96; SPAP | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| whole serum with 0.05% sodium azide | |
| Q13113, Q923S2, Q9CQH0 | |
| PDZK1IP1 | |
| A synthetic peptide made to the C-terminus of mouse MAP17. | |
| RUO | |
| 10158, 67182, 81916 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido