Descripción
- Cubis™ II MCA color touch screen for fast and simple configuration of parameters and workflows.
- Integrated calibration workflows for ABA, ABBA, AB, BnA cycles to ensure efficient, error-free mass comparison.
- Fully integrated function for determining the measurement uncertainty in accordance with OIML and ASTM recommendations.
- Filters for optimal adaptation of the mass comparator to ambient conditions.
- Additional applications for density determination, statistics and individual identifiers are integrated as standard programs.
- Automatic Motorized Leveling.
- Easy logging of reference weight data.
- Continuous weighing range display: any weight between 0 g and the maximum capacity can be displayed
Especificaciones
Especificaciones
| Capacidad | 1110 g |
| Tipo de pantalla | Color Touchscreen |
| Legibilidad | 0,01 mg |
| Tipo | Comparator |