Learn More
Watson-Marlow Manifold Tubing - Silicone
Descripción
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Especificaciones
Especificaciones
| Diámetro interior (imperial) | 3/50 in. |
| Diámetro interior (métrico) | 1,65 mm |
| Material | Silicona |
| Grosor (imperial) | 0.03 in. |
| Grosor (métrico) | 0.8 mm |
| Para utilizar con | 400DM2 and 400DM3 pump heads and multichannel cartridge |
| Tamaño de los tubos | 1,65 mm |
| Caudal | 0.31 to 22 (four roller pump), 0.24 to 17 (eight roller pump)mL/min. |
| Color | Azul/azul |
| Tipo de producto | Pump Tubing |