Descripción
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Conexión | Male Luer Slip, Female Luer Thread |
| Material (sello) | Policarbonato |