Descripción
Easily transition from a luer to thread fitting. Luer fittings allow for a simple yet secure twist-lock connection. NPT thread fittings are designed to create a tight seal under pressure
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Tamaño de rosca | 1/8 27 NPT(macho) |
| Material (sello) | Polipropileno |
| Tipo de producto | Male Luer Integral Lock Ring Hex |