Learn More
Value Plastics Male Luer with Integral Lock Ring to 500 Series Barb
Tienda Cole-Parmer Productos
Click to view available options
Diámetro interior (imperial):
1/16 in
1/4 in
1/8 in
Material:
Nailon
Polipropileno
Tamaño de la unidad:
paquete de 25
Descripción
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;RoHS |
| Diámetro interior (imperial) | 1/4 in |
| Material | Nailon |
| Conexión | Male Luer x Hose Barb |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.