missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAFG Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA540582
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: KRVTQKEELE KQKAELQQEV EKLASENASM KLELDALRSK YEALQTFART Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%; Zebrafish: 93%.
Globin gene expression is regulated through nuclear factor erythroid-2 (NFE2) elements located in enhancer-like locus control regions positioned many kb upstream of alpha- and beta-gene clusters (summarized by Blank et al., 1997 [PubMed 9166829]). NFE2 DNA-binding activity consists of a heterodimer containing a ubiquitous small Maf protein (MafF, MIM 604877; MafG; or MafK, MIM 600197) and the tissue-restricted protein p45 NFE2 (MIM 601490). Both subunits are members of the activator protein-1-like superfamily of basic leucine zipper (bZIP) proteins (see MIM 165160).
Especificaciones
| MAFG | |
| Polyclonal | |
| Unconjugated | |
| MAFG | |
| AA545192; avian musculoaponeurotic fibrosarcoma virus (v-maf) AS42 oncogene, protein G; basic leucine zipper transcription factor MafG; C630022N07Rik; hMAF; MAF bZIP transcription factor G; MAFG; MGC13090; MGC20149; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog G | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17134, 4097 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O15525, O54790 | |
| MAFG | |
| Synthetic peptide directed towards the middle region of human MAFG. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido