Learn More
MADCAM1 Mouse anti-Human, Clone: 314G8, Abnova™
Mouse monoclonal antibody raised against human MADCAM1.
Marca: Abnova MAB12575.100ug
Descripción
The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined. [provided by RefSeq]
Especificaciones
| MADCAM1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA Flow Cytometry (1:10-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user. | |
| MACAM1 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 8174 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
| 314G8 | |
| Mouse monoclonal antibody raised against human MADCAM1. | |
| In PBS, pH 7.4. | |
| MADCAM1 | |
| Recombinant protein corresponding to human MADCAM1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.