missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD2L1BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA565029
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: PDLEWYEKSEE THASQIELLE TSSTQEPLNA SEAFCPRDCM VPVVFPGPVS QEGCCQFTCE LLKHIMYQRQ QLPLPY Highest antigen sequence identity to the following orthologs - mouse 72%, rat 73%.
The protein encoded by this gene was identified as a binding protein of the MAD2 mitotic arrest deficient-like 1 (MAD2/MAD2L1). MAD2 is a key component of the spindle checkpoint that delays the onset of anaphase until all the kinetochores are attached to the spindle. This protein may interact with the spindle checkpoint and coordinate cell cycle events in late mitosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Especificaciones
| MAD2L1BP | |
| Polyclonal | |
| Unconjugated | |
| MAD2L1BP | |
| 0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; caught by MAD2 protein; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1 binding protein; MAD2L1-binding protein; Mad2l1bp; Mad2lbp; mKIAA0110; p31(comet); RP1-261G23.6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9587 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15013 | |
| MAD2L1BP | |
| Recombinant Human MAD2L1BP. Recombinant protein control fragment (Product #RP-104592). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido